Skip to Content
MilliporeSigma
All Photos(12)

Key Documents

ZRB1582

Sigma-Aldrich

Anti-CCPG1 Antibody, clone 1F15 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Cell cycle progression protein 1, Cell cycle progression restoration protein 8

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1F15, monoclonal
recombinant monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 87.34 kDa
observed mol wt ~34 kDa (recombinant fragment)
observed mol wt ~90 kDa

species reactivity

human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... CCPG1(9236)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 1F15 is a ZooMAb rabbit recombinant monoclonal antibody that specifically detects human Cell cycle progression protein 1 (CCPG1). It targets an epitope within the lumenal domain in the C-terminal half.

Immunogen

His-tagged recombinant fragment corresponding to 250 amino acids from the C-terminal, lumenal domain of human Cell cycle progression protein 1 (CCPG1).

Application

Anti-CCPG1, clone 1F15 ZooMAb, Cat. No. ZRB1582, is a recombinant Rabbit monoclonal antibody that specifically targets CCPG1 and is tested for use in Affinity Binding Assay, Immunocytochemistry, Immunohistochemistry, and Western Blotting.
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected CCPG1 in A549 cell lysate.

Affinity Binding Assay: A representative lot of this antibody bound recombinant CCPG1 with a KD of 3.9 x 10-7 in an affinity binding assay.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected CCPG1 in SK-MEl-28 cells.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected CCPG1 in human small intestine tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

Cell cycle progression protein 1 (UniProt: Q9ULG6; also known as Cell cycle progression restoration protein 8, CCPG1) is encoded by the CCPG1 (also known as CCP8, CPR8, KIAA1254) gene (Gene ID: 9236) in human. CCPG1 is a single-pass type II membrane protein that acts as an assembly platform for Rho protein signaling complexes. It is reported to mediate recruitment of endogenous Src kinase into Dbs-containing complexes and interacts with the Rho family member Cdc42. It limits guanine nucleotide exchange activity of MCF2L toward RHOA, which results in an inhibition of both its transcriptional activation ability and its transforming activity. However, it does not inhibit activity of MCF2L toward CDC42, or activity of MCF2 toward either RHOA or CDC42. CCPG1 also serves as an ER stress-inducible endoplasmic reticulum (ER)-phagy cargo receptor in mammals. It is shown to bind directly to ATG8 and FIP200 via distinct peptide motifs. It is reported to offer protection against ER luminal protein aggregation and consequent unfolded protein response hyperactivation and tissue injury of the exocrine pancreas. CCPG1 contains a cytoplasmic domain (aa 1-217), a transmembrane domain (aa 218-238), and a luminal domain (aa 239-757). This ZooMAb recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Smith, MD., et al. (2018). Dev. Cell. 44(2); 217-232; Kostenko EV., et al. (2006). Mol. Cell. Biol. 26(23); 8964-8975).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

300 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service