Skip to Content
MilliporeSigma
All Photos(12)

Key Documents

ZRB1518

Sigma-Aldrich

Anti-Bcl11A Antibody, clone 2C4 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

B-cell CLL/lymphoma 11A, B-cell lymphoma/leukemia 11A, COUP-TF-interacting protein 1, EVI-9, Ecotropic viral integration site 9 protein homolog, Zinc finger protein 856

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit (recombinant)

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

2C4, monoclonal
recombinant monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 91.2 kDa
observed mol wt ~120 kDa

species reactivity

human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... BCL11A(53335)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 2C4 is a ZooMAb rabbit recombinant monoclonal antibody that specifically detects B-cell lymphoma/leukemia 11A (Bcl11A). It targets an epitope within the C-terminal half.

Immunogen

His-tagged recombinant fragment corresponding to 207 amino acids from the internal region of human B-cell lymphoma/leukemia 11A (Bcl11A).

Application

Anti-Bcl11A, clone 2C4 ZooMAb, Cat. No. ZRB1518, is a recombinant Rabbit monoclonal antibody that specifically targets Bcl11A and is tested for use in Affinity Binding Assay, Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting.
Western Blotting Analysis: 1:1,000 dilution from a representative lot detected Bcl11A in Ramos and Jurkat cell lysates.

Affinity Binding Assay: A representative lot of this antibody bound human recombinant Bcl11A with a KD of 2.2 x 10-8 in an affinity binding assay.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected Bcl11A in human tonsil tissue sections.

Immunocytochemistry Analysis: A 1:1,000 dilution from a representative lot detected Bcl11A in HeLa cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

B-cell lymphoma/leukemia 11A (UniProt: Q9H165; also known as BCL-11A, B-cell CLL/lymphoma 11A, COUP-TF-interacting protein 1, Ecotropic viral integration site 9 protein homolog, EVI-9, Zinc finger protein 856) is encoded by the BCL11A (also known as CTIP1, EVI9, KIAA1809, ZNF856) gene (Gene ID: 53335) in human. Bcl11A is a zinc-finger transcription factor that is involved in the generation of B-cell progenitor cells. It contains six C2H2 type zinc finger regions and recognizes TGACCA motif through zinc-finger domain. It is expressed at high levels in brain, spleen thymus, bone marrow and testis. Its expression has also been reported in CD34-positive myeloid precursor cells, B-cells, monocytes and megakaryocytes. Its expression is tightly regulated during B-cell development. Several isoforms of Bcl11A have been reported. In adult erythroid cells, two isoforms are specifically expressed: Isoform 1, also known as Bcl11A-XL, is the more abundant form; Isoform 2, also known as Bcl11A-L, does not have the terminal zinc-finger region. Both these isoforms are predominantly located in the nucleus. Bcl11A associates with the BAF SWI/SNF chromatin remodeling complex. It is involved in hematopoiesis and brain development and is an essential factor in B-cell formation in fetal liver. Chromosomal aberrations involving BCL11A are associated with B-cell malignancies. This ZooMAb recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Liu, N., et al. (2018). Cell 173(2); 430-442).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

0.3 mg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service