Skip to Content
MilliporeSigma
All Photos(15)

Key Documents

ZRB1413

Sigma-Aldrich

Anti-Cryptochrome-1 Antibody, clone 2C22 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Cry1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

recombinant monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 66.4 kDa
observed mol wt ~60 kDa

species reactivity

human

species reactivity (predicted by homology)

bovine, monkey

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
western blot: suitable

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... CRY1(1407)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 2C22 is a ZooMAb rabbit recombinant monoclonal antibody that specifically detects Cryptochrome-1 (CRY1). It targets an epitope within 18 amino acids from the C-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 18 amino acids from the C-terminal region of human Cryptochrome-1 (CRY1).

Application

Anti-Cryptochrome-1, clone 2C22 ZooMAb, Cat. No. ZRB1413, is a recombinant Rabbit monoclonal antibody that targets CRY1 and is tested Affinity Binding Assay, Immunocytochemistry and Western Blotting.
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected Cryptochrome-1 in THP-1 and HeLa cell lysate.

Immunocytochemistry Analysis: A 1:1,000 dilution from a representative lot detected Cryptochrome-1 in HeLa, HUVEC, and A431 cells.

Affinity Binding Assay: A representative lot of this antibody bound CRY1 peptide with a KD of 1.7 x 10-6 in an affinity binding assay.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Cryptochrome-1 (UniProt: Q16526; also known as Cry1) is encoded by the CRY1 (also known as PHLL1) gene (Gene ID: 1407) in human. Cry1 is an evolutionarily conserved member of the flavoprotein superfamily that acts as a transcriptional repressor and forms a core component of the circadian clock that consists of two major components: the central clock, residing in the suprachiasmatic nucleus (SCN) of the brain, and the peripheral clocks that are present in nearly every tissue and organ system. Transcription and translation of core clock components (CLOCK, NPAS2, ARNTL/BMAL1, ARNTL2/BMAL2, PER1, PER2, PER3, CRY1 and CRY2) plays a critical role in rhythm generation. Transcription factors, CLOCK or NPAS2 and ARNTL/BMAL1 or ARNTL2/BMAL2, form the positive limb of the feedback loop and act in the form of a heterodimer and activate the transcription of core clock genes and clock-controlled genes. The core clock genes: PER1/2/3 and CRY1/2 which are transcriptional repressors form the negative limb of the feedback loop and interact with the CLOCK|NPAS2-ARNTL/BMAL1|ARNTL2/BMAL2 heterodimer inhibiting its activity and thereby negatively regulating their own expression. Cry1 and Cry2 are suggested to have redundant functions, but they also differential and selective contributions in defining the pace of the SCN circadian clock and its circadian transcriptional outputs. Cry2 is reported to play a critical role in tuning SCN circadian period by opposing the action of Cry1. Cry is believed to act as a molecular gatekeeper to maintain CLOCK-ARNTL/BMAL1 in a controlled and repressed state until the proper time for transcriptional activation. Cry1 has a photolyase domain at its N-terminal (aa 3-132) and has three FAD biding sites (aa 252, 289, and 355). However, only a minority of the protein molecules contain bound FAD. Cry1 also contains 13 LC3-interacting region (LIR motifs) that are required for its autophagic degradation. A segment of 100 amino acids (371-470) from the C-terminal half is shown to be essential for inhibition of CLOCK-ARNTL/BMAL1-mediated transcription. Animals with defective Cry1 display disruption in their glucose homeostasis with elevated blood glucose in response to acute feeding after an overnight fast and severely impaired glucose clearance in a glucose tolerance test. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Storage and Stability

Recommended storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service