Skip to Content
MilliporeSigma
All Photos(9)

Documents

ZRB1331

Sigma-Aldrich

Anti-PARD-3 Antibody, clone 1N18 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1N18, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 108.54 kDa
observed mol wt ~115 kDa

species reactivity

human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal half

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

lone 1N18 is a is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects Partitioning defective 3 homolog (PARD-3). It targets and epitope within the N-terminal half.

Immunogen

His-tagged recombinant fragment corresponding to 288 amino acids from the N-terminal half of human Partitioning defective 3 homolog (PARD-3).

Application

Quality Control Testing

Evaluated by Western Blotting in U2OS cell lysate.

Western Blotting Analysis: A 1:10,000 dilution of this antibody detected PARD-3 in U2OS cell lysate.

Tested applications

Western Blotting Analysis: A 1:10,000 dilution from a representative lot detected PARD-3 in HeLa cell lysate.

Affinity Binding Assay: A representative lot of this antibody bound PARD-3 with a KD of 1.0 x 10-12 in an affinity binding assay.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected PARD-3 in U2OS cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-PARD-3, clone 1N18 ZooMAb®, Cat. No. ZRB1331, is a recombinant Rabbit monoclonal antibody that specifically targets PARD-3 and is tested for use in Affinity Binding Assay, Immunocytochemistry, and Western Blotting.

Target description

Partitioning defective 3 homolog (UniProt: Q8TEW0; also known as PAR-3, PARD-3, Atypical PKC isotype-specific-interacting protein, ASIP, CTCL tumor antigen se2-5, PAR3-alpha) is encoded by the PARD3 (also known as PAR3, PAR3A) gene (Gene ID: 56288) in human. PARD-3 is an adapter protein that is involved in asymmetrical cell division and cell polarization processes and plays a central role in the formation of epithelial tight junctions. It is localized along the cell-cell contact region. PARD-3 is shown to inhibit atherosclerosis onset by blocking endothelial inflammation. It also colocalizes with the cortical actin that overlays the meiotic spindle during metaphase I and metaphase II. In its N-terminal half, it contains three PDZ domains and a conserved oligomerization domain that is shown to be essential for proper subapical membrane localization. Its second PDZ domain (aa 461-546) mediates interaction with membranes containing phosphoinositol lipids. Together with PARD6, atypical protein kinase C (aPKC), and the Rho GTPase CDC42, it forms a complex that regulates cytoskeleton re-organization, post-Golgi vesicle trafficking, microtubule network polarization, and asymmetric cell division. PARD-3 can undergo phosphorylation at serine 827 by PRKCZ and PRKCI at the most apical tip of epithelial cell-cell contacts during the initial phase of tight junction formation and promote dissociation of the complex with PARD6. Mutations in PARD3 gene cause neural tube defects leading to congenital malformations of the central nervous system. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Hikita, T., et al. (2018). EMBO reports. 19; e45253).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

300 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service