Skip to Content
MilliporeSigma
All Photos(12)

Documents

ZRB13190

Sigma-Aldrich

Anti-BLBP/FABP7 Antibody, clone 7I23, ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

B-FABP, BLBP, Brain lipid-binding protein, Brain-type fatty acid-binding protein, Fatty acid-binding protein, Fatty acid-binding protein 7, Mammary-derived growth inhibitor related

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit (recombinant)

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

7I23, monoclonal
recombinant monoclonal

description

7I23 Clone

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 14.89 kDa
observed mol wt ~15 kDa

species reactivity

rat, human, mouse

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

immunofluorescence: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... FABP7(2173)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 7I23 is a ZooMAb rabbit recombinant monoclonal antibody that specifically detects Brain lipid-binding protein (BLBP).

Immunogen

GST/His-tagged full-length recombinant human Brain lipid-binding protein.

Application

Anti-BLBP/FABP7, clone 7I23 ZooMAb, Cat. No. ZRB13190, is a a recombinant rabbit monoclonal antibody that specifically detects Brain lipid-binding protein (BLBP) and has been tested for use in Immunofluorescence, Immunohistochemistry (Paraffin), and Western Blotting.
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected BLBP/FABP7 in rat brain, human brain, and His-tagged recombinant FABP 2,3,4,7.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected BLBP/FABP7 in human brain tissue sections.

Immunofluorescence Analysis: A 1:100 dilution from a representative lot detected BLBP/FABP7 in human cerebellum tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Fatty acid-binding protein, brain (UniProt: O15540; also known as Brain lipid-binding protein, BLBP, Brain-type fatty acid-binding protein, B-FABP, Fatty acid-binding protein 7, Mammary-derived growth inhibitor related) is encoded by the FABP7 (also known as BLBP, FABPB, MRG) gene (Gene ID: 2173) in human. FABP7 is a member of the large family of hydrophobic ligand-binding proteins (FABPs). FABP proteins are involved in lipid metabolism, including uptake and intracellular trafficking of fatty acids and retinoids. They also play a role in gene regulation, cell signaling, cell growth, and differentiation. FABP7 is a brain-specific protein that is required for the establishment of the radial glial fiber system in the developing brain, a system that is necessary for the migration of immature neurons to establish cortical layers. It is expressed at high levels in the developing central nervous system. It is shown to be important in early nervous system development and is expressed in astrocytes and neuronal cell precursors in mature brain. FABP7 expression is reported to be higher during the light period and lower during the dark in brain regions involved in sleep and activity mechanisms. It is found in melanocytic lesions and has shown to regulate proliferation and invasion in melanoma cells in vitro, suggesting that it may play a role in cell proliferation and invasion. This ZooMAb recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Gerstner, JR et al. (2008). PLoS One 3(2); e1631).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

300 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service