Skip to Content
MilliporeSigma
All Photos(23)

Key Documents

ZRB1274

Sigma-Aldrich

Anti-GSDMD Antibody, clone 1D11 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Gasdermin-D;Gasdermin domain-containing protein 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1D11, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 52.80 kDa
observed mol wt ~50 kDa

purified by

using Protein A

species reactivity

human, mouse

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

epitope sequence

Internal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1D11 is a Rabbit recombinant monoclonal antibody that specifically detects Gasdermin-D. It targets an epitope within 25 amino acids from the internal region.

Immunogen

KLH-conjugated linear peptide corresponding to 25 amino acids from the internal region of human Gasdermin-D.

Application

Quality Control Testing

Evaluated by Western Blotting in A431 cell lysate.

Western Blotting Analysis: A 1:10,000 dilution of this antibody detected GSDMD in A431 cell lysate.

Tested applications

Affinity Binding Assay: A representative lot of this antibody bound GSDMD with a KD of 3.0 x 10-7 in an affinity binding assay.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected GSDMD in human stomach tissue sections.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected GSDMD in A431, HeLa, HUVEC, and NIH3T3 cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

Gasdermin-D (UniProt: P57764; also known as Gasdermin domain-containing protein 1) is encoded by the GSDMD1 (also known as DFNA5L, GSDMDC1, FKSG10) gene (Gene ID: 79792) in human. Gasdermin-D is a member of the gasdermin family of proteins that promotes inflammatory cell death (pyroptosis) in response to microbial infection and danger signals. Its expression is observed in the suprabasal cells of esophagus, as well as in the isthmus/neck, pit, and gland of the stomach. Gasdermin-2 consists of a 31 kDa N-terminal and a 22 kDa C-terminal domain that are separated by a linker region. Under normal conditions, the full-length Gasdermin-D is inactive as the linker loop between the N-terminal and C-terminal domains stabilizes the overall conformation of the full-length protein and allows the C-terminal domain to fold back on and auto-inhibit the N-terminal Gasdermin-D (Gasdermin-N). Upon interdomain cleavage by the action of caspase-1 and 4 at Asp275, the autoinhibition is relieved and Gasdermin-N cytotoxicity is triggered. Gasdermin-N is reported to form membrane pore that lead to cytokine release and pyroptosis. It also displays bactericidal activity. Gasdermin-N released from pyroptotic cells into the extracellular milieu can bind to and kills both Gram-negative and Gram-positive bacteria, without harming neighboring mammalian cells. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Lieberman, J., et al. (2019). Sci. Immunol. 4(39); eaav1447; Liu, X., et al. (2016). Nature. 535(7610); 153-158).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service