Skip to Content
MilliporeSigma
All Photos(14)

Key Documents

ZRB1219

Sigma-Aldrich

Anti-MBP Antibody, clone 2N6 , ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Myelin A1 protein, Myelin basic protein, Myelin membrane encephalitogenic protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit (recombinant)

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

2N6, recombinant monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 17.34 kDa
calculated mol wt 33.11 kDa
observed mol wt 18-20 kDa

species reactivity

mouse, rat, human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... MBP(4155)
mouse ... Mbp(17196)
rat ... Mbp(24547)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 2N6 is a ZooMAb rabbit recombinant monoclonal antibody that specifically detects Myelin Basic Protein (MBP). It targets an epitope within 19 amino acids from the C-terminal half.

Immunogen

KLH-conjugated linear peptide corresponding to 19 amino acids from the C-terminal half of human Myelin Basic Protein.

Application

Anti-MBP, clone 2N6 ZooMAb, Cat. No. ZRB1219, is a highly specific recombinant rabbit monoclonal antibody that detects Myelin basic protein and has been tested for use in Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting.
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected MBP in mouse and rat brain tissue lysate.

Immunocytochemistry Analysis: A 1:10 dilution from a representative lot detected MBP in Rat Glial precursor cells.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected MBP in human brain and mouse brain tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens and experimental conditions may vary.

Target description

Myelin basic protein (UniProt: P02686; also known as MBP, Myelin A1 protein, Myelin membrane encephalitogenic protein) is encoded by the MBP gene (Gene ID: 4155) in human. MBP is a homodimeric protein that is found in both the central and the peripheral nervous system. It is one of the most abundant protein components of the myelin membrane in the CNS and plays a role in both the formation and stabilization of this myelin membrane. It is responsible for adhesion of the cytosolic surfaces of multi-layered compact myelin. MBP can interact with many polyanionic proteins including actin, tubulin, calmodulin, and clathrin, and with negatively charged lipids. MBP isoforms are found in both the central and the peripheral nervous system, whereas Golli-MBP isoforms are expressed in fetal thymus, spleen and spinal cord, as well as in cell lines derived from the immune system. MBP is a major constituent of the myelin sheath of oligodendrocytes and Schwann cells in the nervous system. MBP expression begins abruptly in 14-16-week-old fetuses. Even smaller isoforms seem to be produced during embryogenesis; some of these persisting in the adult. The reduction in the surface charge of citrullinated and/or methylated MBP could result in a weakened attachment to the myelin membrane that can lead to demyelinating diseases such as chronical multiple sclerosis (MS), and fulminating Marburg disease. This ZooMAb recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

30 μg/mL after reconstitution at 25 μL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service