Skip to Content
MilliporeSigma
All Photos(11)

Key Documents

ZMS1229

Sigma-Aldrich

Anti-m3G/TMG cap Antibody, clone K121 ZooMAb® Mouse Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

2,2,7- trimethylguanosine, 2,2,7-TMG cap, m3G

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

mouse

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

K121, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

purified by

using protein G

species reactivity

mouse, human

species reactivity (predicted by homology)

all

packaging

antibody small pack of 25

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunocytochemistry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable

isotype

IgG1κ

greener alternative category

shipped in

ambient

storage temp.

2-8°C

General description

We are committed to bringing you greener alternative products, which adhere to one or more of the 12 Principles of Green Chemistry. This antibody is preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed, and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone K121 is a ZooMAb® mouse recombinant monoclonal antibody that specifically detects m3G/TMG.

Immunogen

KLH-conjugated 2,2,7-trimethylguanosine

Application

Quality Control Testing

Evaluated by Immunocytochemistry in HeLa cells.

Immunocytochemistry Analysis: A 1:100 dilution of this antibody detected m3G/TMG cap in HeLa cells.

Tested Applications

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected m3G/TMG cap in A431, HepG2, and NIH3T3 cells.

Immunofluorescence Analysis: A 1:1,000 dilution from a representative lot detected m3G/TMG cap in Human spleen tissue sections.

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected m3G/TMG cap in Human spleen and Human cervix tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

The 2,2,7-trimethylguanosine (TMG, m3G) cap structure is an important functional characteristic of ncRNAs with critical cellular roles. It is one of the earliest identified RNA modifications that ensures nuclear localization of small nuclear RNAs (snRNAs). It affects pre-mRNA synthesis/splicing, RNA cytoplasmic transport, and mRNA translation and turnover. Shortly after transcription, 7-methylguanosine (m7G), known as cap structure, is added to 5 -end of newly synthesized U snRNA and m7G-capped U snRNA is exported to the cytoplasm. The m7G-cap is converted to m3G-cap, which is a hypermethylated structure of 5 -end, by cytoplasmic catalytic protein TSG1 and the m3G-capped U snRNA forms snRNP with Sm proteins in the cytoplasm, followed by entry into the nucleus. Capping of RNAs is an important requirement for RNA transport and splicing processes. Uncapped RNAs are rapidly degraded by the 5 exoribonuclease enzyme. The TMG-cap also plays a key role in recruiting telomerase to telomeres and engaging Cajal bodies in telomere maintenance. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Yedavalli, YSRK., and Jeang, K-T. (2010). Proc. Natl. Acad. Sci. USA. 107(33); 14787-14792; Moreno, PMD., et al. (2009). Nucleic Acids Res. 37(6); 1925-1935).

Physical form

Purified recombinant mouse monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service