Skip to Content
MilliporeSigma
All Photos(20)

Documents

ZMS1135

Sigma-Aldrich

Anti-Ki-67 Antibody, clone Ki-S5 ZooMAb® Mouse Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Proliferation marker protein Ki-67;Antigen identified by monoclonal antibody Ki-67;Antigen KI-67;Antigen Ki67

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

mouse

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

Ki-S5, recombinant monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 358.69 kDa

purified by

using protein G

species reactivity

human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable

isotype

IgG1

epitope sequence

Unknown

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

mouse ... MKI67(4288)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone Ki-S5 is a ZooMAb® Mouse recombinant monoclonal antibody that specifically detects Ki-67 protein.

Immunogen

Full-length native nuclear protein from human cell line U937.50.

Application

Quality Control Testing

Evaluated by Immunohistochemistry (Paraffin) in human tonsil tissue sections.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution of this antibody detected Ki-67 in human tonsil tissue sections.

Tested Applications

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected Ki-67 in human skin tissue sections.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected Ki-67 in A431, HeLa, and HepG2 cells.

Flow Cytometry Analysis: 0.1 μg from a representative lot detected Ki-67 in one million HeLa cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Proliferation marker protein Ki-67 (UniProt: P46013; also known as Antigen identified by monoclonal antibody Ki-67, KI-67, Ki67) is encoded by the MKI67 gene (Gene ID: 4288) in human. Ki-67 antigen is the prototypic cell cycle related nuclear protein that is expressed by proliferating cells in all phases of the active cell cycle. In the G1 phase it is predominantly localized in the perinucleolar region and in the later phases it is also detected throughout the nuclear interior, being predominantly localized in the nuclear matrix. Its levels are shown to be the highest in the G2 phase and during mitosis. Ki-67 is routinely used as a marker of cell cycling and proliferation. It is required to maintain individual mitotic chromosomes dispersed in the cytoplasm following nuclear envelope disassembly. It is shown to associate with the surface of the mitotic chromosome, the perichromosomal layer, and covers a substantial fraction of the chromosome surface and prevents chromosomes from collapsing into a single chromatin mass by forming a steric and electrostatic charge barrier. Ki67 binds DNA, with a preference for supercoiled DNA and AT-rich DNA. It undergoes phosphorylation and is hyperphosphorylated in the mitosis phase. The hyperphosphorylated form fails to bind DNA. Ki-67 contains an N-terminal forkhead-associated (FHA) domain (aa 27-76), a protein phosphatase 1 (PP1) binding domain (aa 502-549), a large central region comprising 16 tandem repeats, and a C-terminal leucine/arginine-rich chromatin-binding (LR) domain. Ki-67 interacts with kinesin-like motor protein Kif15 and a nucleolar protein NIFK (Nucleolar Protein Interacting with the FHA domain of MKI67) via its FHA domain to promote cell proliferation and tumor metastasis. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Sun, X., and Kaufman PD. (2018). Chromosoma. 127(2); 175-186; MacCallum, DE., and Hall, PA. ( 2000). J. Pathol. 191(3); 286-298).

Physical form

Purified recombinant mouse monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

30 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service