Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z763993

StripSpin 12 mini centrifuge adapter

fits 0.5 mL tubes, pack of 8 ea

Synonym(s):

strip spin adapter, mini centrifuge adapter

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103911
NACRES:
NB.22

feature

Holds 0.5 mL tubes

packaging

pack of 8 ea

manufacturer/tradename

Benchmark Scientific C1008-A5-8

technique(s)

centrifugation: suitable

compatibility

for use with MC-12 High Speed Microcentrifuge
for use with MC-24 High Speed Microcentrifuge
for use with myFuge 12 Mini Centrifuge
for use with myFuge 5 MicroCentrifuge
for use with myFuge Mini Centrifuge


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service