Key Documents
Z757810
Sartorius filter pipette tips
volume range 50-1000 μL, sterile, pack of 960 ea (10 racks of 96 tips)
Synonym(s):
filter pipette tips, pipetting tips, pipettor tips
About This Item
UNSPSC Code:
41121601
NACRES:
NB.01
volume:
50—1000 μL
sterility:
sterile
feature:
DNase free
RNase free
barrier
filter
packaging:
pack of 960 ea (10 racks of 96 tips)
manufacturer/tradename:
Sartorius 791001F
material
colorless
colorless tip
polypropylene (Virgin)
sterility
sterile
feature
DNase free
RNase free
barrier
filter
packaging
pack of 960 ea (10 racks of 96 tips)
manufacturer/tradename
Sartorius 791001F
maximum volume
1 mL
volume range
50-1000 μL
volume
50—1000 μL
binding type
low binding surface
General description
The Sartorius filter tips of the Biohitfamily are made of polypropylene and feature a polyethylene filter barrier to prevent aerosol and liquid contamination. This filter helps protect against the rist of cross-contamination and reduces maintenance requirements of the pipette. The filter tips are suited for molecular biology, microbiology, cell culture and radioactive work.
These tips have more space between the sample and the filter than conventional filter tips so you do not need to worry about the sample touching the filter whatever the pipetting technique or type of liquid.
Benefits:
  • Pipette foaming liquids like buffers and proteins
  • Pipette solvents
  • Reverse pipetting
  • Multiple dispensing functions of electronic pipettes
Other Notes
Browse all pipette tips
Choose from one of the most recent versions:
Certificates of Analysis (COA)