Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z744265

Wilmad® NMR Tube Carrier with Shirt Clip

to hold, 1 NMR tube, tube O.D. 5 mm, tube L 7 in., pack of 3 ea (pack of 3)

Synonym(s):

NMR tube carrier, NMR tube holder

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121705
NACRES:
NB.15

feature

to hold 1 NMR tube

packaging

pack of 3 ea (pack of 3)

manufacturer/tradename

Wilmad WG-6192

tube L

7 in.

tube O.D.

5 mm

General description

Handy pocket NMR/EPR tube carrier holds one 5mm O.D., 7 inch length NMR/EPR tube. Pack of 3 white holders. Features a pocket clip that keeps it secure. Made from unbreakable polymer material with a snap-on cap. Protection is provided for the sample and lab personnel during transport.

Application

NMR

Legal Information

Wilmad is a registered trademark of SP Industries

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service