Key Documents
Z743507
Direct Thermal WetGrip labels
roll of 1000
About This Item
UNSPSC Code:
55121611
NACRES:
NB.35
packaging
roll of 1000
manufacturer/tradename
Diversified Biotech GRDT-3000
L × W
1.50 in. × 0.75 in.
General description
GRIPS - To wet, frost-covered and frozen surfaces; temperature resistant to -80°C
THIN - Compatible with centrifuge rotors
RESISTANT - To chemicals and solvents
MULTI - SURFACE - Adheres to plastics, glass, metals and even paperboard boxes
For use with the GoDEX DT200 Direct printer and Dymo LabelWriter 450 (DYMO-550 printer is NOT compatible)
Disclaimer
Important note for International Customers: Our Direct Thermal labels are designed to work with U.S. Model Dymo 450 printers only. They are not compatible with European Dymo printers or Dymo 550 printers.