MilliporeSigma
Z742001
Low Profile Roller
Lab Start-Up Kit
Synonym(s):
Laboratory roller, Low-profile roller
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NB.22
UNSPSC Code:
41103811
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
AC/DC input
94 V / 240 V AC (Power Cords - 115V US and 230V European)
feature
Includes Five Replacement Rollers (Z367737) and bag of assorted o-rings
manufacturer/tradename
IBI ROLLS0001
platform size
12 in. × 12 in.
General description
The IBI Low Profile Roller is a lightweight unit designed as a compact, space saving, non-dedicated instrument. This unit will operate on a benchtop, inside high humidity and CO2incubators, high temperature ovens (60°C), and refrigerators (0°C). The IBI Low Profile Roller comes standard with six rollers in place for rotating three large cell culture bottles. The versatility of this unit allows the user to add up to five additional rollers quickly and easily. No tools are needed to move or add rollers. Moving rollers closer together or adding more rollers allows this system to roll small tubes from 15mL to 50mL. Rubber O-Rings to be placed on the small tubes are included with every unit.
The low profile design provides a convenient fit into laboratory incubators, ovens, and refrigerated units. The thin, flat 115V power cord fits easily through incubator doors without compromising the environment inside. An industrial grade chain drive provides years of trouble-free service. All product finishes are resistant to chemicals and easy to clean and sterilize. The rollers can be easily removed for simple cleaning and adjusting to accommodate different tube sizes. The cool-running, brushless DC motor creates no ozone, and the universal input voltage allows the unit to operate at 90VAC to 240VAC. The roller provides a wide operational temperature range (0-60°C), and the ability to operate in high humidity (up to 97%) and CO2.
The Low Profile Roller Lab Start-Up Kit comes with eleven rollers for rolling up to 20 small tubes ranging in size from 15 to 50mL. Each Lab Start-Up Kit comes with a bag of assorted O-rings to be placed on the tubes to ensure they are level and remain in place on the roller.
The Low Profile Roller was previously manufactured by Stovall Scientific. All IBI accessories and replacement parts are compatible with the Stovall Low Profile Roller.
Still not finding the right product?
Explore all of our products under Low Profile Roller
Choose from one of the most recent versions:
Certificates of Analysis (COA)