Key Documents
Z741772
Sigma-Aldrich
Sigma Aldrich® Dual Run & Blot Accessory
Sponge Pad Kit, size 9 cm × 9 cm, pack of 4 ea
About This Item
UNSPSC Code:
41100000
NACRES:
NB.22
packaging
pack of 4 ea
manufacturer/tradename
Sigma-Aldrich
size
9 cm × 9 cm
General description
Sigma Aldrich gels run best on our Dual Run and Blot unit. This unique apparatus permits the running of both 10x10cm and 8x10cm gels. The unit is available in dual and quad form enabling up to 4 gels to be run at one time.
Both units can also be used to complete Western blots. The unique electrode arrangement provides a gradient electric field which ensures that both small and large proteins migrate from the gel at a comparable rate.
Application
Sigma Aldrich Sponge Pad Kit consists of four foam pads for use when blotting with Sigma Aldrich Blotting Cassettes.
Legal Information
SIGMA-ALDRICH is a registered trademark of Merck KGaA, Darmstadt, Germany