Key Documents
Z741669
Sartorius VALUEPAK pipette tips
volume range 0.1-10 μL
About This Item
UNSPSC Code:
41121600
NACRES:
NB.01
manufacturer/tradename:
Sartorius 790012MAX
maximum volume:
10 μL
volume range:
0.1-10 μL
sterility:
non-sterile
feature:
DNase free
RNase free
barrier: no
endotoxin free
packaging:
pack of 960 ea (Refil)
pkg of (refill Tower)
material
colorless
sterility
non-sterile
feature
DNase free
RNase free
barrier: no
endotoxin free
packaging
pack of 960 ea (Refil)
pkg of (refill Tower)
manufacturer/tradename
Sartorius 790012MAX
maximum volume
10 μL
volume range
0.1-10 μL
General description
Large value packs of tips with the perfect tip fitting and sealing to the pipetter. This gives you the best possible accuracy and precision at the best possible price. Compatible with Optiload feature in Biohit Picus, eLINE®, mLINE® and Proline® Plus pipetters, enabling ergonomic and light tip attachment and ejection.
Other Notes
Browse all pipette tips
Legal Information
Proline is a registered trademark of Sartorius Biohit Liquid Handling Oy
eLINE is a registered trademark of Sartorius Biohit Liquid Handling Oy
mLINE is a registered trademark of Sartorius Biohit Liquid Handling Oy
Choose from one of the most recent versions:
Certificates of Analysis (COA)