Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z740271

Aluminum heating/cooling block

Holds 192 x 0.2 mL tubes in center plus 20 on size (8 for 1.5ml tubes, 12 for 0.5ml tubes)

Synonym(s):

dry block, tube cooling block, tube heating block

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41102404
NACRES:
NB.35
Pricing and availability is not currently available.

material

aluminum

feature

Holds 192 x 0.2 mL tubes in center plus 20 on size (8 for 1.5ml tubes, 12 for 0.5ml tubes)

packaging

pkg of 1 ea

manufacturer/tradename

Diversified Biotech CHAM-1020

technique(s)

PCR: suitable

size

156x171x25 mm

Looking for similar products? Visit Product Comparison Guide

General description

Each Cooling Chamber is a solid aluminum block that surrounds inserted tubes to guarantee sustained hot or cold temperatures.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service