Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z728594

Hettich® adapter for 50 mL conical tubes

for ROTANTA 460/460R Centrifuge, Adapter for 5 x 50 mL Conical Tubes 29 x 115 mm

Synonym(s):

Hettich® centrifuge adapters

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103911
NACRES:
NB.22

feature

Holds 50 mL tubes

packaging

pack of 1 ea

manufacturer/tradename

Hettich 4740

technique(s)

centrifugation: suitable

compatibility

for use with 5625-A

Looking for similar products? Visit Product Comparison Guide

Related Categories

Legal Information

Hettich is a registered trademark of Andreas Hettich GmbH & Co. KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service