Documents
Z722731
Hettich® MIKRO 220/220R centrifuge accessories
for MIKRO 220/220R Centrifuge, Swing-out Rotor, 24 place (w/lid)
About This Item
UNSPSC Code:
41103900
NACRES:
NB.22
feature
lid
to hold 24 x 1.5/2.0 mL tubes
packaging
pack of 1 ea
manufacturer/tradename
Hettich 1154-L
parameter
18,516 max. RCF
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models MIKRO 220/220R
Legal Information
Hettich is a registered trademark of Andreas Hettich GmbH & Co. KG