Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

Z720895

Nunc® Cell Factory, Nunclon® Δ surface

10-chamber CF, 6320 cm2, 2000 mL, 6/cs

Synonym(s):

cell culture bioreactor, cell culture chamber, cell factories

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122104
NACRES:
NB.22

material

polystyrene

sterility

sterile

packaging

pack of 1
case of 6

manufacturer/tradename

Nunc 170009

L × W

335mm × 205 mm

surface area

6320 cm2

working volume

2000 mL

color

clear

suitability

suitable for (industrial scale cell culture; production of biomaterials such as vaccines, monoclonal antibodies, etc.)

Looking for similar products? Visit Product Comparison Guide

General description

Nunc® Cell Factory, Nunclon® Δ surface

  • For large scale cell culture and production of biomaterials such as vaccines, monoclonal antibodies, etc
  • Ideal for adherent cells
  • Can be used for suspension culture as well
  • Growth kinetics unaltered from laboratory scale culture
  • Available in 1, 2, 4, 10 and 40 tray cell factory versions for easy scale-up
  • Compact design, large growth surface in limited space areas
  • Low risk of contamination
  • Easy to handle multi-layer cell factory stacks
  • Polystyrene trays assembled by sonic welding one on another
  • Not Autoclavable
  • Certified Nunclon® Δ surface treatment ensures excellent conditions for cell attachment and growth
  • Each unit is individually wrapped twice

Legal Information

Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Nunclon is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hye Jin Shin et al.
Viruses, 13(2) (2021-02-07)
Genetic screens using CRISPR/Cas9 have been exploited to discover host-virus interactions. These screens have identified viral dependencies on host proteins during their life cycle and potential antiviral strategies. The acyl-CoA binding domain containing 3 (ACBD3) was identified as an essential

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service