Key Documents
Z710997
Wheaton media bottle replacement cap
rubber lined cap only (size 33-430)
Synonym(s):
240280
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
manufacturer/tradename
WHEATON®
Features and Benefits
  • Rubber lined caps feature a nontoxic white styrene-butadiene rubber liner ideal for cell culture work
Legal Information
WHEATON is a registered trademark of Wheaton Industries, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)