Key Documents
Z694126
Tygon® 2475 high purity tubing
About This Item
UNSPSC Code:
41105108
NACRES:
NB.35
packaging
pack of 50 feet
manufacturer/tradename
Saint Gobain ACG00017
I.D. × O.D.
1/4 in. × 3/8 in. × 1/16 in.
General description
TYGON® 2475 high-purity tubing is a plasticizer-free tubing that utilizes the latest in polymer technology to provide an entirely clear and flexible tubing choice for sensitive fluid transfer applications. TYGON 2475 is hydrophobic and will resist the absorption/adsorption of aqueous fluids. This reduction in sorption minimizes the risk of fluid alteration in single- or repeat-use applications. The smooth inner surface of TYGON 2475 inhibits particulate entrapment on the tubing wall and reduces the potential for bacterial growth and contamination.
Features and Benefits
  • Exceptionally low absorption/adsorption compared to silicone
  • Smooth inner surface inhibits particulate entrapment
  • Environmentally safe to dispose and reduces disposal costs
  • Highly resistant to aggressive cleaners and sanitizers
  • Plasticizer-free
  • Meets all USP Class VI and FDA criteria
Typical Applications:
  • Sterile filling and dispensing systems
  • Diagnostic equipment
  • Laboratory analytical instrumentation
  • Infusion sets for parenterals and drugs
  • Cell and tissue culture transport
Legal Information
Tygon is a registered trademark of Saint-Gobain Performance Plastics Corp.
Choose from one of the most recent versions:
Certificates of Analysis (COA)