Key Documents
Z333913
BRAND® accu-jet® pro pipette controller membrane filters
spare membrane filter 0.2 μm, non-sterile
Synonym(s):
bottle top filters, laboratory filters, vacuum system filters
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
Pricing and availability is not currently available.
sterility
non-sterile
packaging
pkg of 10 ea
manufacturer/tradename
BRAND 26535
General description
The accu-jet®S / pro / macro pipette controller is designed to safeguard your equipment with its robust PP housing and integrated interior membrane that effectively blocks liquid. This protective feature ensures the longevity of your pipette controller while maintaining a clean and safe working environment.
Other Notes
BR26535 (India)
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
accu-jet is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)