Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

X3003

Sigma-Aldrich

Xanthosine 3′,5′-cyclic monophosphate

≥98% (HPLC), solid

Synonym(s):

cXMP

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

Quality Level

Assay

≥98% (HPLC)

form

solid

mol wt

apparent mol wt 367.2

packaging

vial of 2 mg

storage temp.

−20°C

SMILES string

OC1C2OP(O)(=O)OCC2OC1n3cnc4C(=O)NC(=O)Nc34

InChI

1S/C10H11N4O8P/c15-5-6-3(1-20-23(18,19)22-6)21-9(5)14-2-11-4-7(14)12-10(17)13-8(4)16/h2-3,5-6,9,15H,1H2,(H,18,19)(H2,12,13,16,17)

InChI key

AAUQFLOPCIXUSK-UHFFFAOYSA-N

Biochem/physiol Actions

cXMP is a an analog of cyclic GMP that is used to evaluate phosphodiesterase activity.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Moss et al.
The Journal of biological chemistry, 252(15), 5211-5215 (1977-08-10)
Guanosine 3':5'-monophosphate phosphodiesterases, which appear to be under allosteric control, have been partially purified from rat liver supernatant and particulate fractions. The preferred substrate for both phosphodiesterases was cGMP (Km values: cGMP less than cIMP less than cAMP). At subsaturating

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service