MilliporeSigma
WPB01603
Whirl-Pak® Thio-Bag®, Waste Water Sampling Bag
capacity 1000 mL (34 oz), bag W × L × thickness 15 cm × 38 cm × 0.102 mm
Synonym(s):
Nasco B01603, sample bag, sample collection, sample receptacle, sample storage bag
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NB.35
UNSPSC Code:
24111500
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
material
clear linear low-density polyethylene (LLDPE)
sterility
sterile
feature
label plain white
packaging
pkg of 600 ea (6 boxes of 100EA)
manufacturer/tradename
Whirl-Pak® B01603
bag W × L × thickness
15 cm × 38 cm × 0.102 mm
capacity
1000 mL (34 oz)
General description
Meets National Environmental Laboratory Accreditation Conference (NELAC) standard, June 2003.
Capacity is measured in fluid ounces when the bag is closed and tab is folded over three times. Volume and dimensions are approximate; bags should not be used at temperatures above 180° F (82° C). Bags can be frozen to any temperature, even liquid nitrogen temperatures down to -346° F (-210° C) - careful handling required after freezing. All bags are sterilized after manufacturing.
Features and Benefits
  • EPA approved
  • Self-standing gusseted bag
  • Sodium thiosulfate tablet in the bag for chlorine neutralization
  • Built-in puncture proof closure
  • Post-manufacturing sterilization for results you can trust.
Legal Information
Thio-Bag is a registered trademark of The Aristotle Corporation
Whirl-pak is a registered trademark of The Aristotle Corporation
Still not finding the right product?
Choose from one of the most recent versions:
Certificates of Analysis (COA)