Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

WHA10416306

Whatman® Nytran SuPerCharge (SPC) TurboBlotter refills - small

Nytran SPC 0.45 μm TurboBlotter refill, 11 x 14 cm, 1/pk

Synonym(s):

Z381950, blotting membrane, nylon membrane

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105300
NACRES:
NB.22

packaging

pack of 1

manufacturer/tradename

Whatman 10416306
Whatman Article No. 28415384 (US reference)

size

11 cm × 14 cm

suitability

suitable for (high-resolution transfer of DNA and RNA; nucleic acid transfer)

Looking for similar products? Visit Product Comparison Guide

General description

TurboBlotter Refills, Small
Nytran SPC, wicks and blotter stacks for five transfers from indicated gel size.

Note: Replacement packet of Turboblotter consumables, sufficient for 5 transfers.

Features and Benefits

  • Rapid: Downward capillary transfer allows for alkaline buffer transfers in 1 h and neutral (SSC) transfers in 3 h.
  • Economical: Reusable blotting device requires less buffer and blotting paper. Convenient refill packs available.
  • Compact: Has a smaller footprint than most homemade devices and is stackable. Up to five units can be stacked on top of each other during transfers.
  • Easy to use: Very easy to set up and works without power or vacuum source.
  • Uses Whatman Nytran SPC membrane and Whatman 3MM Chr and GB003 or GB005 blotting papers for top performance.

Other Notes

Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.

Legal Information

Nytran is a trademark of Cytiva
Whatman is a registered trademark of Cytiva

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service