Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

WH0255631M7

Sigma-Aldrich

Monoclonal Anti-COL24A1 antibody produced in mouse

clone 1D8, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-collagen, type XXIV, alpha 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
ELISA (i)
WB
clone:
1D8, monoclonal
species reactivity:
human
citations:
2
technique(s):
indirect ELISA: suitable
western blot: 1-5 μg/mL

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1D8, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

Collagen type XXIV α 1 (COL24A1) is part of the fibril-forming family of collagen molecules and is expressed in bones.

Immunogen

COL24A1 (NP_690850, 1626 a.a. ~ 1714 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
PRWTSTQTSGPGLPIGFKGWNGQIFKVNTLLEPKVLSDDCKIQDGSWHKATFLFHTQEPNQLPVIEVQKLPHLKTERKYYIDSSSVCFL

Biochem/physiol Actions

Collagen type XXIV α 1 (COL24A1) takes part in modulating various processes in the cartilage and bone. It has been linked to head and neck squamous cell carcinoma.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

CREB-AP1 protein complexes regulate transcription of the collagen XXIV gene (Col24a1) in osteoblasts.
Matsuo N
The Journal of Biological Chemistry, 281(9), 5445-5452 (2006)
Kiyoshi Misawa et al.
Molecular and clinical oncology, 2(2), 285-291 (2014-03-22)
Squamous cell carcinoma of the head and neck (HNSCC) is the sixth most common type of cancer, affecting ~500,000 individuals worldwide annually. Collagen is the major constituent of the extracellular matrix component in tumors and plays a crucial role in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service