Skip to Content
MilliporeSigma
All Photos(5)

Key Documents

WH0055669M4

Sigma-Aldrich

Monoclonal Anti-MFN1 antibody produced in mouse

clone 3C9, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-DKFZp762F247, Anti-FLJ20693, Anti-MGC41806, Anti-hfzo1, Anti-hfzo2, Anti-mitofusin 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3C9, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MFN1(55669)

General description

The protein encoded by this gene is a mediator of mitochondrial fusion. This protein and mitofusin 2 are homologs of the Drosophila protein fuzzy onion (Fzo). They are mitochondrial membrane proteins that interact with each other to facilitate mitochondrial targeting. (provided by RefSeq)

Immunogen

MFN1 (AAH40557, 1 a.a. ~ 741 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MAEPVSPLKHFVLAKKAITAIFDQLLEFVTEGSHFVEATYKNPELDRIATEDDLVEMQGYKDKLSIIGEVLSRRHMKVAFFGRTSSGKSSVINAMLWDKVLPSGIGHITNCFLSVEGTDGDKAYLMTEGSDEKKSVKTVNQLAHALHMDKDLKAGCLVRVFWPKAKCALLRDDLVLVDSPGTDVTTELDSWIDKFCLDADVFVLVANSESTLMNTEKHFFHKVNERLSKPNIFILNNRWDASASEPEYMEDVRRQHMERCLHFLVEELKVVNALEAQNRIFFVSAKEVLSARKQKAQGMPESGVALAEGFHARLQEFQNFEQIFEECISQSAVKTKFEQHTIRAKQILATVKNIMDSVNLAAEDKRHYSVEEREDQIDRLDFIRNQMNLLTLDVKKKIKEVTEEVANKVSCAMTDEICRLSVLVDEFCSEFHPNPDVLKIYKSELNKHIEDGMGRNLADRCTDEVNALVLQTQQEIIENLKPLLPAGIQDKLHTLIPCKKFDLSYNLNYHKLCSDFQEDIVFRFSLGWSSLVHRFLGPRNAQRVLLGLSEPIFQLPRSLASTPTAPTTPATPDNASQEELMITLVTGLASVTSRTSMGIIIVGGVIWKTIGWKLLSVSLTMYGALYLYERLSWTTHAKERAFKQQFVNYATEKLRMIVSSTSANCSHQVKQQIATTFARLCQQVDITQKQLEEEIARLPKEIDQLEKIQNNSKLLRNKAVQLENELENFTKQFLPSSNEES

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elodie Lobet et al.
Scientific reports, 8(1), 5173-5173 (2018-03-28)
Mitochondria are complex organelles that participate in many cellular functions, ranging from ATP production to immune responses against viruses and bacteria. This integration of a plethora of functions within a single organelle makes mitochondria a very attractive target to manipulate
Maria Kawalec et al.
Folia neuropathologica, 52(4), 436-442 (2015-01-13)
Mitofusin 2 (Mfn2), a protein of the mitochondrial outer membrane, is essential for mitochondrial fusion and contributes to the maintenance and operation of the mitochondrial network. Mutations in the mitofusin 2 gene cause axonal Charcot-Marie-Tooth type 2A (CMT2A), an inherited
Kayleen Vannuvel et al.
Journal of cellular physiology, 231(9), 1913-1931 (2015-12-19)
Endoplasmic reticulum (ER) and mitochondria are not discrete intracellular organelles but establish close physical and functional interactions involved in several biological processes including mitochondrial bioenergetics, calcium homeostasis, lipid synthesis, and the regulation of apoptotic cell death pathways. As many cell
Lilian Phu et al.
Molecular cell, 77(5), 1107-1123 (2020-03-07)
Mitochondria import nearly their entire proteome from the cytoplasm by translocating precursor proteins through the translocase of the outer membrane (TOM) complex. Here, we show dynamic regulation of mitochondrial import by the ubiquitin system. Acute pharmacological inhibition or genetic ablation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service