Skip to Content
MilliporeSigma
All Photos(8)

Key Documents

WH0006591M5

Sigma-Aldrich

Monoclonal Anti-SNAI2 antibody produced in mouse

clone 3C12, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-MGC10182, Anti-SLUG, Anti-SLUGH1, Anti-WS2D, Anti-snail homolog 2 (Drosophila)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3C12, monoclonal

form

buffered aqueous solution

species reactivity

mouse, rat, human

technique(s)

indirect ELISA: suitable
indirect immunofluorescence: suitable
western blot: 1-5 μg/mL

isotype

IgG3κ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SNAI2(6591)

Related Categories

General description

This gene encodes a member of the Snail family of C2H2-type zinc finger transcription factors. The encoded protein acts as a transcriptional repressor that binds to E-box motifs and is also likely to repress E-cadherin transcription in breast carcinoma. This protein is involved in epithelial-mesenchymal transitions and has antiapoptotic activity. Mutations in this gene may be associated with sporatic cases of neural tube defects. (provided by RefSeq)

Immunogen

SNAI2 (NP_003059, 97 a.a. ~ 169 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
KDHSGSESPISDEEERLQSKLSDPHAIEAEKFQCNLCNKTYSTFSGLAKHKQLHCDAQSRKSFSCKYCDKEYV

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wu Zhou et al.
Journal of cell science, 126(Pt 12), 2740-2746 (2013-04-18)
Epithelial-mesenchymal transition (EMT) can be induced by several pleiotropically activated transcription factors, including the zinc-finger E-box-binding protein Zeb1. Mechanisms regulating Zeb1 expression have been partly uncovered, showing a critical role for the miR-200 family members. In the present study, we
Gaohua Han et al.
International journal of molecular medicine, 39(2), 357-363 (2016-12-31)
The aim of this study was to examine the effect of Annexin A1 (ANXA1) on the proliferation, migration and invasion of esophageal squamous cell carcinoma (ESCC) cells and its possible mechanisms of action. After constructing the ANXA1 overexpression plasmid, we transfected this plasmid
Damian Ryszawy et al.
Carcinogenesis, 35(9), 1920-1930 (2014-02-08)
Suppressive function of connexin(Cx)43 in carcinogenesis was recently contested by reports that showed a multifaceted function of Cx43 in cancer progression. These studies did not attempt to model the dynamics of intratumoral heterogeneity involved in the metastatic cascade. An unorthodox

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service