Skip to Content
MilliporeSigma

W0515

Well orienter

96 wells

Synonym(s):

Diversified Biotech WELL-1000, microplate well orienter, numbered well orienter, pipetting aid, well orienter plate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.22
UNSPSC Code:
41122102
Manufacturer/tradename:
Diversified Biotech WELL-1000
Packaging:
pack of 4 ea
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


sterility

non-sterile

packaging

pack of 4 ea

manufacturer/tradename

Diversified Biotech WELL-1000

size

96 wells

color

none

General description

A perfectly sized plastic card with sharp, alpha-numeric characters to identify each and every well on a microplate, will end cross referencing headaches. Once positioned under the microplate, the Well Orienter makes identifying sample wells easy and headache-free!


Still not finding the right product?

Explore all of our products under Well orienter




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library