Skip to Content
MilliporeSigma
All Photos(1)

Documents

V4762

Sigma-Aldrich

Monoclonal Anti-Vascular Endothelial Growth Factor Receptor-1 antibody produced in mouse

clone FLT-19, tissue culture supernatant

Synonym(s):

Anti-Flt1 Receptor, Anti-VEGF R-1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

tissue culture supernatant

antibody product type

primary antibodies

clone

FLT-19, monoclonal

contains

15 mM sodium azide

species reactivity

human

technique(s)

immunohistochemistry (frozen sections): 1:100 using human placenta
immunoprecipitation (IP): suitable
indirect ELISA: suitable
microarray: suitable

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FLT1(2321)

General description

Monoclonal Anti-VEGF Receptor-1 (Flt1 Receptor) (mouse IgG1 isotype) is derived from the FLT-19 hybridoma produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with a recombinant extracellular domain of VEGF Receptor-1 (Flt1 Receptor) of human origin. Vascular endothelial growth factor (VEGF), also called vasculotropin (VAS)1 and vascular permeability factor (VPF) 2 is a member of a family of endothelial cell mitogens and angiogenic factors. VEGF is a homodimeric heparin-binding glycoprotein. VEGF binds to two structurally similar receptor tyrosine kinases; Flt15 (fms-like tyrosine kinase 1, also termed VEGF Receptor-1, VEGF-R1) and KDR6 (kinase-insert domain containing receptor, also termed VEGF-R2). Flt1 is predominately expressed in human placenta and human vascular endothelial cells, while KDR is more widely expressed in all vessel-derived endothelial cells but low in human and fetal bovine placenta.
Mouse monoclonal clone FLT-19 anti-Vascular Endothelial Growth Factor Receptor-1 antibody recognizes the extracellular domain of human VEGF Receptor-1 molecule (Flt1 Receptor). The antibody does not recognize VEGF Receptor-2 (KDR), VEGF receptor-3 (sFlt4,) and PDGF-Rβ.

Immunogen

recombinant human extracellular domain of VEGFR-1.

Application

Monoclonal Anti-Vascular Endothelial Growth Factor Receptor-1 antibody produced in mouse has been used in:
  • immunofluorescence labelling
  • enzyme-linked immunosorbent assay (ELISA)
  • immunohistochemistry
  • immunoprecipitation

Mouse monoclonal clone FLT-19 anti-Vascular Endothelial Growth Factor Receptor-1 antibody may be used for the localization of VEGF-R1 using various immunochemical assays such as ELISA, immunoblotting, immunocytochemistry, immunohistochemistry, and immunoprecipitation. Antibodies that react specifically with VEGF receptors are useful for the study of the specific differential tissue expression and intracellular localization of the receptor in normal and neoplastic tissue.

Biochem/physiol Actions

The mitogenic activity of VEGF appears to be stimulated by specific VEGF receptors (160-200 kDa) which can be found on the surface of various endothelial cells. VEGF binds to two structurally similar receptor tyrosine kinases; Flt1 (fms-like tyrosine kinase 1, also known as VEGF Receptor-1 (VEGF-R1), and KDR (kinase insert domain containing receptor, also known as VEGF-R2). Studies using KDR and Flt1 stably transfected endothelial cell lines have shown that these two receptors exhibit different affinities to VEGF and mediate different responses. KDR/Flk1 does not respond to placental growth factor (PlGF), a VEGF related growth factor, while Flt1 binds PlGF specifically. Flt1 is predominately expressed in human placenta and human vascular endothelial cells. Both VEGF receptors (KDR and Flt1) are upregulated in human fetal and adult kidney.
Vascular endothelial growth factor (VEGF) stimulates the proliferation of endothelial cells isolated from both small and large vessels.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Real-time ligand binding of fluorescent VEGF-A isoforms that discriminate between VEGFR2 and NRP1 in living cells
PeachnCJ, et al.
Cell Chemical Biology, 25(10), 1208- 1218 (2018)
Sophie Le Ricousse-Roussanne et al.
Cardiovascular research, 62(1), 176-184 (2004-03-17)
Recent studies have provided increasing evidence that postnatal neovascularization does not rely exclusively on sprouting of preexisting vessels, but also involves bone marrow-derived circulating endothelial precursors (BM-EPCs). Animal studies revealed that neovascularization of ischemic tissue can be enhanced by BM-EPCs
Vascular endothelial cell growth factor (VEGF), an emerging target for cancer chemotherapy
Shinkaruk S, et al.
Current Medicinal Chemistry. Anticancer Agents, 3(2), 95-117 (2003)
A Sawano et al.
Blood, 97(3), 785-791 (2001-02-07)
Flt-1, also known as vascular endothelial growth factor receptor 1 (VEGFR-1), is a high-affinity tyrosine kinase receptor for VEGF and is expressed almost exclusively on vascular endothelial cells. As an exception, Flt-1 transcript was recently found to be expressed in
Chloe J Peach et al.
Cell chemical biology, 25(10), 1208-1218 (2018-07-31)
Fluorescent VEGF-A isoforms have been evaluated for their ability to discriminate between VEGFR2 and NRP1 in real-time ligand binding studies in live cells using BRET. To enable this, we synthesized single-site (N-terminal cysteine) labeled versions of VEGF165a, VEGF165b, and VEGF121a.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service