Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

U5260

Sigma-Aldrich

UNISETTE Biopsy Processing/Embedding Cassettes

white

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116124
NACRES:
NB.17

color

white

application(s)

hematology
histology

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

General description

Histology processing/embedding cassettes made of acetal polymer. These cassettes keep specimens safely submerged in liquid and are resistant to histological solvents. The one-piece lid snaps apart and conveniently locks into the cassette base, ensuring specimen safety. A line of cassette colors are available to fit your needs and preference. A large 30° anterior writing area is available for your convenience. Not suitable for automated printers.

Application

Compatible with Leica and Sakura printers.

Quantity

Case contains 3 dispenser boxes of 500 cassettes.

Legal Information

Unisette is a trademark of Simport

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Walid Ellouze et al.
Canadian journal of microbiology, 64(8), 527-536 (2018-04-11)
Understanding the variation in how wheat genotypes shape their arbuscular mycorrhizal (AM) fungal communities in a prairie environment is foundational to breeding for enhanced AM fungi-wheat interactions. The AM fungal communities associated with 32 durum wheat genotypes were described by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service