Skip to Content
MilliporeSigma

TMO375418

Nunc® cryogenic tubes

polypropylene, capacity 1.8 mL, round bottom, starfoot, external thread, sterile, case of 1800 ea

Synonym(s):

biobanking tube, cell culture cryogenic tube, Nunc 375418, cryogenic sample storage, cryogenic tube

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.43
UNSPSC Code:
41121704
Material:
polypropylene , round bottom
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

polypropylene , round bottom

sterility

sterile

feature

Writing Surface, A-R Base

packaging

case of 1800 ea

manufacturer/tradename

Nunc 375418

technique(s)

cryopreservation: suitable

capacity

1.8 mL

General description

Nunc®cryogenic tubes are designed for the storage of samples, ranging from general cold storage to the vapor phase of liquid nitrogen. These tubes are available with either internal or external threads to suit different preferences and requirements.

Features and Benefits

  • Non-cytotoxic, non-mutagenic raw materials and pyrogen-free IVD compatible tubes for added sample security.
  • Certified SAL of 10-6 for the highest sterility assurance available.
  • Suitable for storage from general cold storage refrigeration (+4°C) down to the vapor phase of liquid nitrogen.

Legal Information

Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries


Still not finding the right product?

Explore all of our products under Nunc® cryogenic tubes




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library