Skip to Content
MilliporeSigma

TMO236707

Nunc® sealing tape for 96-well plate

(Polyester), non-sterile, case of 1000 ea

Synonym(s):

adhesive film dispenser, adhesive tape dispenser, tape dispenser, Nunc 236707, sealing film

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.15
UNSPSC Code:
41161502
Manufacturer/tradename:
Nunc 236707
Material:
(Polyester)
Sterility:
non-sterile
Packaging:
case of 1000 ea
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

(Polyester)

sterility

non-sterile

packaging

case of 1000 ea

manufacturer/tradename

Nunc 236707

General description

Minimize evaporation and protect samples from contamination and spilling with Thermo Scientific Nunc Sealing Tapes. These adhesive seals are ideal for all assay applications. Nunc Life Science products offer reliability and reproducible results, and are backed by industry-leading quality standards and certifications.

Application

  • General purpose
  • ELISA

Features and Benefits

  • Adhesive seals effectively seal all microplate formats.
  • Pressure sensitive adhesive is not tacky to touch, but forms a strong bond to the plate upon application of pressure.
  • Working temperature -40°C to +120°C
  • Non sterile.
  • Not autoclavable.
  • .

Legal Information

Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library