Skip to Content
MilliporeSigma

Skip To

T9525

Tris-Borate-EDTA buffer

working solution

Synonym(s):

TBE buffer

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.25
PubChem Substance ID:
UNSPSC Code:
41105319
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


sterility

0.2 μm filtered

Quality Segment

form

working solution

technique(s)

electrophoresis: suitable

impurities

DNase, RNase, Protease, none detected

suitability

suitable for gel electrophoresis (after dilution to working concentration)

application(s)

diagnostic assay manufacturing

SMILES string

OB(O)O.NC(CO)(CO)CO.OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O

InChI

1S/C10H16N2O8.C4H11NO3.BH3O3/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20;5-4(1-6,2-7)3-8;2-1(3)4/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20);6-8H,1-3,5H2;2-4H

InChI key

OSBLTNPMIGYQGY-UHFFFAOYSA-N

Application

TBE running buffer is the most commonly used buffer for DNA and RNA polyacrylamide gel electrophoresis. TBE is used with non-denaturing or denaturing (7 M urea) gels. It is also routinely used for DNA automated sequencing gel. TBE can also be used for agarose gels but is not recommended for preparative gels for recovery of nucleic acids.
Dilution of the TBE stock concentrates to a 1× TBE running buffer results in a buffer containing 89 mM Tris-borate and 2 mM EDTA, pH 8.3. The 5× or 10× stocks may also be added to an acrylamide/bis-acrylamide stock solution for making the PAGE gel. Applied voltages of less than 5 V/cm (distance between the electrodes of the unit) are recommended for maximum resolution.

Packaging

Supplied in dispenser with a spigot.

Preparation Note

Prepared with Biotechnology Performance Certified Trizma base (Product Code T6066) and Molecular Biology Reagents boric acid (Product Code B6768) and EDTA disodium salt (Product Code E5134).
Solution prepared with 18 megohm water

Other Notes

89 mM Tris borate, pH approx. 8.3, containing 2 mM EDTA.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
T6025T3913T4415
technique(s)

electrophoresis: suitable

technique(s)

electrophoresis: suitable

technique(s)

electrophoresis: suitable

technique(s)

-

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

application(s)

diagnostic assay manufacturing

application(s)

diagnostic assay manufacturing

application(s)

diagnostic assay manufacturing

application(s)

-

form

working solution

form

working solution

form

powder blend

form

solution

suitability

suitable for gel electrophoresis (after dilution to working concentration)

suitability

-

suitability

suitable for gel electrophoresis (after dilution to working concentration)

suitability

-

sterility

0.2 μm filtered

sterility

0.2 μm filtered

sterility

-

sterility

-


Still not finding the right product?

Explore all of our products under Tris-Borate-EDTA buffer


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Repr. 1B

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters