Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T9429

Sigma-Aldrich

Anti-Transforming Growth Factor-β, pan antibody produced in rabbit

IgG fraction of antiserum, lyophilized powder

Synonym(s):

Anti-TGF-β, pan

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

amphibian, human, pig

technique(s)

microarray: suitable
neutralization: suitable
western blot: suitable

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Specificity

The antibody will neutralize the biological activity of human TGF-β1, porcine TGF-β1.2, porcine TGF-β2, human TGF-β3, and amphibian TGF-β5.

Immunogen

mixture of recombinant human TGF-β1, porcine TGF-β1.2, porcine TGF-β2, and recombinant amphibian TGF-β5.

Biochem/physiol Actions

The effect of intraperitoneal injection of Anti-Transforming Growth Factor-β, pan antibody produced in rabbit in monocrotaline-induced pulmonary hypertension was studied in rat model.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline containing carbohydrates.

Preparation Note

Purified using protein A.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Miao Jia et al.
Frontiers in physiology, 13, 778479-778479 (2022-03-22)
Peritoneal dialysis (PD) is an important part of replacement therapy for kidney failure. However, long-term PD treatment can cause peritoneal fibrosis. Autophagy may be involved in the pathological mechanism of peritoneal fibrosis (PF). Although autophagy is currently known to be
Ya-Wen Hsiao et al.
Journal of immunology (Baltimore, Md. : 1950), 172(3), 1508-1514 (2004-01-22)
IL-6 is a multifunctional cytokine that regulates cell growth, differentiation, and cell survival. Many tumor cells produce TGF-beta1, which allows them to evade CTL-mediated immune responses. IL-6 antagonizes TGF-beta1 inhibition of CD3 cell activation. However, whether IL-6 restores NK activity
Steven M Driever et al.
Philosophical transactions of the Royal Society of London. Series B, Biological sciences, 372(1730) (2017-08-16)
To meet the growing demand for food, substantial improvements in yields are needed. This is particularly the case for wheat, where global yield has stagnated in recent years. Increasing photosynthesis has been identified as a primary target to achieve yield
Aikaterini J Megalou et al.
International journal of clinical and experimental medicine, 3(4), 332-340 (2010-11-13)
The role of transforming growth factor-β in the pathogenesis of pulmonary arterial hypertension is unclear. We examined the effects of T9429, an antibody against transforming growth factor-β receptors, on hemodynamic, histological and functional parameters in the rat model of monocrotaline-induced
Tal Fisher et al.
Cell reports, 39(11), 110954-110954 (2022-06-08)
Severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) leads to shutoff of protein synthesis, and nsp1, a central shutoff factor in coronaviruses, inhibits cellular mRNA translation. However, the diverse molecular mechanisms employed by nsp1 as well as its functional importance are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service