Skip to Content
MilliporeSigma

T9253

Trypsin inhibitor

powder

Synonym(s):

Ovomucoid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.77
EC Number:
232-906-9
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Trypsin inhibitor from chicken egg white, Type II-O, Partially purified ovomucoid, containing ovoinhibitor

biological source

chicken egg white

Quality Segment

type

Type II-O

form

powder

mol wt

28 kDa

solubility

water: soluble 10 g/L

storage temp.

2-8°C

General description

Ovomucoid or trypsin inhibitor is an abundant protein in most avian egg whites. The hen′s egg protein is composed of about 186 amino acid residues, and is highly glycosylated. Three tandem domains are each homologous with pancreatic trypsin inhibitor. It is highly immunogenic, and probably accounts for most cases of egg allergy.
25% of ovomucoid comprises carbohydrate.

Application

Trypsin inhibitor from chicken egg white has been used:
  • to produce denatured, oxidized and deglycosylated ovomucoid and used to stimulate patient derived cell cultures for mapping T cell epitopes
  • to treat nuclei for flow cytometric analysis
  • in Holtfreter′s solution, used for the dissociation of planarians into cells

Analysis Note

One mg will inhibit 0.8-1.6 mg of tryspin with activity of approx. 10,000 BAEE units per mg protein. May inhibit ≤0.3 mg of chymotrypsin with activity of approx. 40 BTEE units per mg protein.

Other Notes

One trypsin unit will produce a ΔA253 of 0.001 per min with BAEE as substrate at pH 7.6 at 25 °C; reaction volume 3.2 mL, 1 cm light path.
Ovomucoid, from chicken egg white, does not itself inhibit chymotrypsin. Chymotrypsin inhibition is a measure of ovoinhibitor contamination by method of Feeney, R. E., et al., J. Biol. Chem., 238, 1415 (1963).
View more information on Trypsin Inhibitor.


Still not finding the right product?


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library