Skip to Content
MilliporeSigma
All Photos(2)

Documents

T8948

Sigma-Aldrich

1-(α,α,α-Trifluoro-m-tolyl)piperazine hydrochloride

99%

Synonym(s):

N-[3-(Trifluoromethyl)phenyl]piperazine hydrochloride, TFMPP hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H13F3N2 · HCl
CAS Number:
Molecular Weight:
266.69
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

99%

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada

availability

not available in Japan

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

239-241 °C (lit.)

solubility

methanol:glacial acetic acid (1:1): soluble 25 mg/mL

application(s)

forensics and toxicology
veterinary

SMILES string

Cl.FC(F)(F)c1cccc(c1)N2CCNCC2

InChI

1S/C11H13F3N2.ClH/c12-11(13,14)9-2-1-3-10(8-9)16-6-4-15-5-7-16;/h1-3,8,15H,4-7H2;1H

InChI key

DGNLGWJZZZOYPT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Preparation Note

1-(α,α,α-Trifluoro-m-tolyl)piperazine hydrochloride is soluble in methanol:glacial acetic acid (1:1) at 25 mg/ml.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

STOT SE 3

Target Organs

Central nervous system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anne Zwartsen et al.
Neurotoxicology, 66, 87-97 (2018-03-25)
While the prevalence and the use of new psychoactive substances (NPS) is steadily increasing, data on pharmacological, toxicological and clinical effects is limited. Considering the large number of NPS available, there is a clear need for efficient in vitro screening
Ushtana Antia et al.
Journal of forensic sciences, 55(5), 1311-1318 (2010-06-11)
An LC-MS method was developed for benzylpiperazine (BZP) and trifluoromethylphenylpiperazine (TFMPP), constituents of "party pills" or "legal herbal highs," and their metabolites in human blood plasma. Compounds were resolved using a mixture of ammonium formate (pH 4.5, 0.01 M) and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service