Skip to Content
MilliporeSigma
All Photos(4)

Documents

T5130

Sigma-Aldrich

TAPS

≥99.5% (titration)

Synonym(s):

N-[Tris(hydroxymethyl)methyl]-3-aminopropanesulfonic acid, [(2-Hydroxy-1,1-bis(hydroxymethyl)ethyl)amino]-1-propanesulfonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H17NO6S
CAS Number:
Molecular Weight:
243.28
Beilstein:
2452420
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥99.5% (titration)

form

crystalline powder

useful pH range

7.7-9.1

pKa 

8.4

mp

230-235 °C (dec.)

solubility

water: 0.5 g/mL, clear, colorless

application(s)

diagnostic assay manufacturing

SMILES string

OCC(CO)(CO)NCCCS(O)(=O)=O

InChI

1S/C7H17NO6S/c9-4-7(5-10,6-11)8-2-1-3-15(12,13)14/h8-11H,1-6H2,(H,12,13,14)

InChI key

YNLCVAQJIKOXER-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service