MilliporeSigma
T1668
Nalgene® centrifuge tubes, Oak Ridge Style 3118
nominal capacity 28 mL
Synonym(s):
High-Speed ​​Centrifuge Tubes, Nalgene tubes, Nalgene 3118-0028, sample collection tube, sample receptacle, sample tube
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NB.11
UNSPSC Code:
41121703
Material:
polypropylene screw closure, round bottom tube, screw top cap, transparent polycarbonate
Packaging:
pack of 10 ea
Feature:
RCF 50,000 × g
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
material
polypropylene screw closure, round bottom tube, screw top cap, transparent polycarbonate
feature
RCF 50,000 × g
packaging
pack of 10 ea
manufacturer/tradename
Nalgene® 3118-0028
nominal capacity
28 mL
General description
“Nalgene” Oak Ridge high-speed polycarbonate centrifuge tubes offer outstanding mechanical strength, allowing for a wide range of applications. Additionally, their transparent nature facilitates easy observation of the contents in the tube.
Features and Benefits
  • Capable of withstanding temperatures ranging from -135°C to +135°C (-211°F to +275°F).
  • Designed with a round bottom to accommodate high-speed rotors.
  • Suitable for use in both refrigerated and non-refrigerated centrifuges.
  • Includes polypropylene linerless closures, which are easy to clean and provide leakproof protection under normal conditions up to 10,000 x g.
  • Autoclavable.
Other Notes
  • Polycarbonate centrifuge tubes will have their lifespan reduced with repeated autoclaving.
  • It is essential to fill the tubes to a minimum of 80% of their total capacity to ensure proper performance.
  • To achieve leakproof performance exceeding 10,000 x g, utilize a sealing cap or sealing cap assembly.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Still not finding the right product?
Choose from one of the most recent versions:
Certificates of Analysis (COA)