MilliporeSigma
T1168
Nalgene® centrifuge tubes, Oak Ridge Style 3119
nominal capacity 28 mL
Synonym(s):
High-Speed ​​Centrifuge Tubes, Nalgene tubes, Nalgene 3119-0028, sample collection tube, sample receptacle, sample tube
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NB.11
UNSPSC Code:
41121703
Material:
polypropylene copolymer , polypropylene screw closure, round bottom tube, screw top cap (Linerless)
Packaging:
pack of 10 ea
Feature:
RCF 50,000 × g
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
material
polypropylene copolymer , polypropylene screw closure, round bottom tube, screw top cap (Linerless)
feature
RCF 50,000 × g
packaging
pack of 10 ea
manufacturer/tradename
Nalgene® 3119-0028
nominal capacity
28 mL
General description
“Nalgene” Oak Ridge High-Speed PPCO Centrifuge Tubes demonstrate superior chemical compatibility with a broader range of laboratory reagents compared to regular polycarbonate centrifuge tubes. Additionally, these PPCO tubes can be repeatedly autoclaved, ensuring prolonged and reliable performance.
Features and Benefits
  • Exhibits higher chemical resistance compared to polycarbonate tubes.
  • Can withstand repeated autoclaving, resulting in a longer lifespan compared to regular polycarbonate tubes subjected to the same conditions.
  • Includes polypropylene linerless closures, which are easy to clean and provide leakproof protection under normal conditions up to 10,000 x g.
  • The tubes are autoclavable and become leakproof when used with the provided Nalgene closure.
Other Notes
  • Tubes should be filled to a minimum of 80% capacity to ensure proper performance.
  • These centrifuge tubes are recyclable in numerous communities.
  • When the product reaches the end of its life, kindly wash and recycle your Nalgene PP and PPCO centrifuge tubes if recycling services are available in your area.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Still not finding the right product?
Choose from one of the most recent versions:
Certificates of Analysis (COA)