Skip to Content
MilliporeSigma

T1135

TERGITOL

MIN FOAM 1x

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
NACRES:
NA.25
PubChem Substance ID:
UNSPSC Code:
12161900
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

description

non-ionic

form

liquid

mol wt

640 g/mol

CMC

34 ppm

transition temp

cloud point 40 °C

solubility

soluble (Water solubility: 10 g/l at 20 °C - soluble)

HLB

12.6

application(s)


SMILES string

OCCO.CCCCO.CC(O)CO

InChI

1S/C4H10O.C3H8O2.C2H6O2/c1-2-3-4-5;1-3(5)2-4;3-1-2-4/h5H,2-4H2,1H3;3-5H,2H2,1H3;3-4H,1-2H2

InChI key

QMNOIORHZMRPLS-UHFFFAOYSA-N

General description

Tergitol is a nonionic surfactant which can be used to extract estrogens

Application

Tergitol has been used in a study to assess a method for detecting Salmonella enterica serovar Enteritidis in liquid whole eggs using loop-mediated isothermal amplification (LAMP). It has also been used in a study to investigate the oxidation of nonylphenol ethoxylates (NPEO) and octylphenol ethoxylates (OPEO) by oxidant systems generating hydroxide radicals.

Other Notes

Polyglycol ether (nonionic) surfactants.

Legal Information

TERGITOL is a trademark of The Dow Chemical Company ("Dow") or an affiliated company of Dow


Storage Class

10 - Combustible liquids

wgk

WGK 2

flash_point_f

392.0 °F - open cup - (External MSDS)

flash_point_c

200 °C - open cup - (External MSDS)

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library