Key Documents
T1043
Sigma-Aldrich
Nalgene® centrifuge tubes, Oak Ridge Style 3119
nominal capacity 10 mL
Synonym(s):
High-Speed ​​Centrifuge Tubes, Nalgene tubes
About This Item
UNSPSC Code:
41121703
NACRES:
NB.11
Pricing and availability is not currently available.
material
polypropylene copolymer
polypropylene screw closure
round bottom tube
screw top cap (Linerless)
Agency
suitable for DIN 38407-42
sterility
non-sterile
feature
RCF 50,000 × g
packaging
pack of 10 ea
manufacturer/tradename
Nalgene® 3119-0010
nominal capacity
10 mL
General description
“Nalgene” Oak Ridge high-speed polypropylene copolymer (PPCO) centrifuge tubes offer broader chemical compatibility with lab reagents compared to polycarbonate centrifuge tubes.
Features and Benefits
  • Provides superior chemical resistance compared to polycarbonate tubes.
  • Autoclavable and can withstand repeated autoclaving, resulting in a longer lifespan than polycarbonate tubes subjected to the same conditions.
  • Includes polypropylene linerless closures, which are easy to clean and provide leakproof protection under normal conditions up to 10,000 x g.
  • Leakproof when used with the provided Nalgene closure.
Other Notes
Fill the tubes to a minimum of 80% capacity to ensure proper performance.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)