Skip to Content
MilliporeSigma

T0691

Dounce tissue grinder tube

working volume 15 mL

Synonym(s):

885303-0015

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.22
UNSPSC Code:
41121800
Material:
glass
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

glass

packaging

pack of 1 ea

working volume

15 mL

General description

Designed primarily for cellular work where the nucleus remains intact after homogenization.

Features and Benefits

  • Manufactured from borosilicate glass 3.3
  • Designed primarily for cellular work where the nucleus remains intact after homogenization.
  • All-glass construction
  • Two pestles are supplied with each complete unit
  • Large clearance pestle is used for the initial sample reduction
  • Small clearance pestle is used to form the final homogenate
  • Replacement components are available and completely interchangeable



Still not finding the right product?

Explore all of our products under Dounce tissue grinder tube




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library