Skip to Content
MilliporeSigma

T0197

Nunc® sealing tape for multiwell plates

aluminum sealing tape, silicone adhesive, pierceable

Synonym(s):

multiwell plate sealing films, sealing films, Nunc 276014, laboratory sealing film, plate seal

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.22
UNSPSC Code:
41122410
Manufacturer/tradename:
Nunc 276014
Material:
aluminum film
Sterility:
non-sterile
Packaging:
pack of 100 ea
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

aluminum film

description

certified RNase/DNase-free

sterility

non-sterile

feature

piercable

packaging

pack of 100 ea

manufacturer/tradename

Nunc 276014

color

silver

suitability

suitable for (general use, PCR, storage, photo-sensitive samples)

General description

A complete line of sealing tapes for use with MicroWell plates in a wide range of applications. The performance of these tapes has been characterized under a variety of conditions, including an extensive study of the evaporation of well contents and the adherence of tapes to plates manufactured in various polymers.

Featuring 3M adhesive technology, sealing tapes provide a highly effective seal, preventing evaporation and eliminating contamination.

Specific tapes are intended for use in General Assays, Genomics, Compound and Library Storage, Bio-Analytical Assays, High Throughput Screening and Drug Discovery applications.

Pierceable with probes or pipet tips without coring

Application

  • For storage, PCR, microscopy, culture, protection

Features and Benefits

  • Provide an effective seal on polypropylene, polystyrene, and polycarbonate microwell plates
  • Compatible with aqueous solutions and organic solvents
  • Tested for use between -80 and +110°C
  • Pre-cut, plate-sized sheets
  • Protective backing
  • Minimal evaporation of plate contents
  • Eliminates well-to-well cross contamination
  • Easy to handle delivery system
  • Provided with removable tabs

Legal Information

Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
T0197-100EA04061834381495