Skip to Content
MilliporeSigma

SYNB135124

Synthware Desiccator with lid and ceramic plate

bottle O.D. 240 mm, clear borosilicate glass 3.3

Synonym(s):

Synthware B135124, desiccator with Top Knob

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.43
UNSPSC Code:
41121800
Material:
clear borosilicate glass 3.3
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

clear borosilicate glass 3.3

packaging

pack of 1 ea

manufacturer/tradename

Kemtech B135124

bottle O.D.

240 mm

General description

Desiccator, Clear, Id 240mm, Includes Lid & Ceramic Plate

Application

kemetch; synthware; desiccator

Features and Benefits

Organic Chemistry

Legal Information

Synthware is a trademark of Kemtech America Inc.


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library