Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP6557

Sigma-Aldrich

Thrombin Active from human plasma

≥95% (SDS-PAGE), recombinant, lyophilized

Synonym(s):

Activated Factor IIa

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

Product Name

Thrombin Active from human plasma, ≥95% (SDS-PAGE)

biological source

human

Assay

≥95% (SDS-PAGE)

form

lyophilized

potency

>3000 units/mg

mol wt

37 kDa

packaging

pkg of 1000 units
pkg of 250 units

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... F2(2147)

General description

Thrombin enzyme (activated factor IIa) is an important clotting promoter that controls the transformation of soluble fibrinogen to insoluble active fibrin strands. Thrombin is a coagulation protein and a serine protease (EC 3.4.21.5) that catalyzes many coagulation-related reactions. It is primarily associated with blood clotting.

Biochem/physiol Actions

Thrombin triggers factor-XI, factor-V, Factor-XIII and factor-VIII. Thrombin endorses platelet activation, using activation of protease-activated receptors on the platelet. It also induces inflammation. As a result of its high proteolytic specificity, thrombin has become an important biochemical protein. The thrombin cleavage site (Leu-Val-Pro-Arg-Gly-Ser) is widely used in linker regions of recombinant fusion protein constructs. After the purification of the fusion protein, thrombin is used to cleave between the Arginine and Glycine residues of the cleavage site, efficiently removing the purification tag from the protein of interest with a high degree of specificity.

Physical form

Lyophilized from a buffer composed of 20 mM Bis-Tris, 150 mM NaCl and 0.1% PEG 8000, pH 6.5.

Reconstitution

A working stock solution can be prepared by adding 1 mL buffered saline, pH 7.4 (ex. 25 mM HEPES, 150 mM NaCl, 0.1% PEG 8000, pH 7.4). Further dilution should be made in buffer containing a suitable stabilizing agent such as 0.1%-1% Prionex, BSA, or PEG.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structure of the thrombin complex with triabin, a lipocalin-like exosite-binding inhibitor derived from a triatomine bug.
Fuentes-Prior P
Proceedings of the National Academy of Sciences of the USA, 94(22), 11845-11850 (1997)
Exogenous thrombin delivery promotes collateral capillary arterialization and tissue reperfusion in the murine spinotrapezius muscle ischemia model.
Bruce AC and Peirce SM
Microcirculation (New York, N.Y. : 1994), 19(2), 143-154 (2012)
Ajjampura C Vinayaka et al.
Pharmaceuticals (Basel, Switzerland), 15(12) (2022-12-24)
Lung inflammation is associated with elevated pro-inflammatory cytokines and chemokines. Treatment with FCBD:std (standard mix of cannabidiol [CBD], cannabigerol [CBG] and tetrahydrocannabivarin [THCV]) leads to a marked reduction in the inflammation of alveolar epithelial cells, but not in macrophages. In
Tobias Strittmatter et al.
Nature chemical biology, 18(10), 1125-1134 (2022-08-09)
Cellular therapies remain constrained by the limited availability of sensors for disease markers. Here we present an integrated target-to-receptor pipeline for constructing a customizable advanced modular bispecific extracellular receptor (AMBER) that combines our generalized extracellular molecule sensor (GEMS) system with
Hadar Peeri et al.
Cancers, 13(7) (2021-05-01)
Glioblastoma multiforme (GBM) is the most lethal subtype of glioma. Cannabis sativa is used for the treatment of various medical conditions. Around 150 phytocannabinoids have been identified in C. sativa, among them Δ-9-tetrahydrocannabinol (THC) and cannabidiol (CBD) that trigger GBM

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service