SRP4171
IL-15 from mouse
recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)
Synonym(s):
IL 15, IL-T, Interleukin 15
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
biological source
mouse
recombinant
expressed in E. coli
Assay
≥98% (HPLC)
≥98% (SDS-PAGE)
form
lyophilized
mol wt
~13.3 kDa
packaging
pkg of 10 μg
storage condition
avoid repeated freeze/thaw cycles
impurities
endotoxin, tested
NCBI accession no.
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
Gene Information
mouse ... Il15(16168)
General description
IL15 (interleukin 15) protein is a short-chain α-helical bundle type 1 T cell-stimulating cytokine. It is a pleiotropic cytokine belonging to the cytokine receptor γ chain family, and is expressed by dendritic cells (DCs), epithelial cells, macrophages, monocytes, and fibroblasts. IL15, along with its receptor IL15Rα, are co-expressed majorly by activated monocytes and DCs (dendritic cells). Recombinant mouse IL-15 is a 13.3 kDa protein containing 115 amino acid residues.
Biochem/physiol Actions
IL15 (interleukin 15) and IL2 are the only two known cytokine till date that bind to T-cells through IL-2Rβ (IL2 receptorβ). It interacts with lymphocytes via the heterotrimeric IL-15 receptor (IL-15R) composed of IL-15Rα, IL-2 receptor beta (IL-2Rβ), and IL-2 receptor gamma (IL-2Rγ). Since, IL-2R and IL-15R have common subunits, it is supposed that IL2 and IL15 perform similar functions, such as induction of lymphocyte activation and proliferation in vitro, and apoptosis of activated lymphocytes (AICD). However, unlike IL2, IL15 mimics the function of IFN I (interferon) by potently and selectively activating memory-phenotype CD44hi CD8+ cells.
Physical form
Lyophilized from 10 mM Tris, pH 8.0.
Reconstitution
Centrifuge the vial prior to opening. Avoid freeze-thaw cycles.
Reconstitute in water to a concentration of 0.1-1.0 μg/μL. The solution can then be diluted into other aqueous buffers.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Interleukin-15 Constrains Mucosal T Helper 17 Cell Generation: Influence of Mononuclear Phagocytes.
PLoS ONE, 10(11) (2015)
Nature communications, 5, 5150-5150 (2014-10-07)
Following microbial pathogen invasion, one of the main challenges for the host is to rapidly control pathogen spreading to avoid vital tissue damage. Here we report that an effector CD8(+) T-cell population that expresses the marker NK1.1 undergoes delayed contraction
Cutting edge: requirement for IL-15 in the generation of primary and memory antigen-specific CD8 T cells.
Journal of Immunology, 168(10), 4827-4831 (2002)
The shared and contrasting roles of IL2 and IL15 in the life and death of normal and neoplastic lymphocytes: implications for cancer therapy.
Cancer immunology research, 3(3), 219-227 (2015)
European journal of immunology, 44(9), 2638-2647 (2014-07-06)
Mature NK cells are heterogeneous as to their expression levels of cell surface molecules. However, the functional differences and physiological roles of each NK-cell subset are not fully understood. In this study, we report that based on the Ly6C expression
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service