Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP3325

Sigma-Aldrich

Uterglobin Peptide

≥98% (HPLC), human recombinant, expressed in E. coli

Synonym(s):

Clara cell phospholipid-binding protein, Secretoglobin family 1A member 1, Urinary protein 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

Product Name

UTERGLOBULIN human, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)

biological source

human

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

mol wt

8.0 kDa

packaging

pkg of 50 μg

impurities

endotoxin, tested

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... SCGB1A1(7356)

General description

Uteroglobin, also known as clara cell phospholipid-binding protein or CC10, is a member of the secretoglobin superfamily. It is secreted from nonciliated cells in the epithelial lining of bronchioles, also referred as Clara cells. Uteroglobin is encoded by the gene mapped to human chromosome 11q12.2 – 13.1. Recombinant Human Uteroglobin is an 8.0kDa homodimeric protein consisting of 142 amino acid residues.

Biochem/physiol Actions

Uteroglobin functions as an immunomodulatory and anti-inflammatory protein. Uteroglobin might play a vital role in regulation of airway inflammation. Elevated level of uteroglobin has been observed in fallopian tubes of tubal ectopic pregnant women and in female patients with pelvic inflammatory disease. Uteroglobin level is reduced in serum of asthmatic patients with a long duration of the disease. Uteroglobin stimulates the growth of preimplantation blastocysts and it functions as a carrier and/or scavenger of progesterone, decreasing its toxicity to the early developing embryo. Uteroglobin has the capacity to prevent both chemotaxis and phagocytosis of monocytes and neutrophils.

Physical form

Lyophilized from 1 x PBS

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Uteroglobin: a novel cytokine?
Mukherjee AB
Cellular and Molecular Life Sciences, 55(5), 771-787 (1999)
Serum levels of Clara cell 10-kDa protein are decreased in patients with asthma.
Shijubo N
Lung, 177(1), 45-52 (1999)
Anil B Mukherjee et al.
Endocrine reviews, 28(7), 707-725 (2007-10-06)
Blastokinin or uteroglobin (UG) is a steroid-inducible, evolutionarily conserved, secreted protein that has been extensively studied from the standpoint of its structure and molecular biology. However, the physiological function(s) of UG still remains elusive. Isolated from the uterus of rabbits
Current knowledge of the aetiology of human tubal ectopic pregnancy.
Shaw JL
Human Reproduction Update, 16(4), 432-444 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service