Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NA.32
UNSPSC Code:
12352202
biological source
human
recombinant
expressed in CHO cells
assay
≥95% (HPLC), ≥95% (SDS-PAGE)
form
lyophilized
mol wt
18.9 kDa
packaging
pkg of 20 μg
impurities
<0.1 EU/μg endotoxin, tested
color
white to off-white
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... IL24(11009)
General description
IL-24 is produced by a variety of cell types, including B cells, CD4+ cells, NK cells, lymph node DCs, monocytes, and melanoma cells. IL-24 can signal through the IL20R1/IL20R2 and IL22R1/IL20R2 receptors to initiate a signaling cascade which includes the induction of JAK1/STAT3 phosphorylation. IL-24 is, functionally, a pleiotropic protein but is generally characterized as an anti-cancer cytokine that can selectively inhibit growth of a wide variety of human cancer cells through activities that include the induction of differentiation and apoptosis, and the suppression of angiogenesis and cell proliferation.
Biochem/physiol Actions
IL-24 is a secreted glycoprotein belonging to the IL-10 structural family of cytokines. Recombinant human IL-24 is an 18.9 kDa glycoprotein, containing 161 amino acid residues, including a C-terminal His-tag.
Physical form
Lyophilized from 10 mM Sodium Phosphate, pH 7.5.
Preparation Note
Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.
Storage Class
10 - Combustible liquids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.